Gene Transcripts

Gene Symbol
Eya2
Entrez Gene ID
156826
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
Chromosome Location
Chr. 3: 162285279 - 162470642 on Build Rnor_6.0
UniGene ID
Rn.39687
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130427.1 NP_569111.1 13 1358
XM_008762456.1 XP_008760678.1 14 1594
GenBank mRNA AY366465.1 - 13 1358

View Full Product Details