Gene Transcripts

Gene Symbol
THAP1
Entrez Gene ID
55145
Gene Name
THAP domain containing 1
Gene Aliases
DYT6
Chromosome Location
Chr. 8: 42836674 - 42843331 on Build GRCh38
UniGene ID
Hs.7432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018105.2 NP_060575.1 3 578
NM_199003.1 NP_945354.1 2 382
GenBank mRNA AK001339.1 - 3 572
AK223231.1 - 3 557
AL832077.1 - 2 356
BC021721.2 - 3 552 4795229
BG497522.1 - 3 579
CD639319.1 - 2 364
CR457256.1 - 3 341

View Full Product Details