Gene Transcripts

Gene Symbol
Thegl
Entrez Gene ID
498350
Gene Name
theg spermatid protein-like
Gene Aliases
Chromosome Location
Chr. 14: 33389511 - 33431883 on Build Rnor_6.0
UniGene ID
Rn.122806
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017498.1 NP_001017498.1 5 956
GenBank mRNA BC087065.1 - 5 956

View Full Product Details