Gene Transcripts

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256532.1 NP_001243461.1 2, 3 306
NM_001256533.1 NP_001243462.1 2, 3 306
NM_018116.3 NP_060586.2 2, 3 306
NR_046292.1 2, 3 306
NR_046293.1 2, 3 306
NR_046294.1 1, 2 398
NR_046295.1 2, 3 306
XM_006711421.1 XP_006711484.1 1, 2 143
XM_006711423.2 XP_006711486.1 2, 3 250
XM_011509695.1 XP_011507997.1 2, 3 264
XM_011509696.1 XP_011507998.1 2, 3 264
XM_011509697.1 XP_011507999.1 2, 3 264
XM_011509698.1 XP_011508000.1 2, 3 264
XM_011509699.1 XP_011508001.1 2, 3 264
XM_011509700.1 XP_011508002.1 2, 3 264
XM_011509701.1 XP_011508003.1 2, 3 264
XM_011509702.1 XP_011508004.1 2, 3 264
XM_011509706.1 XP_011508008.1 2, 3 238
XM_011509709.2 XP_011508011.1 2, 3 264
XM_017001603.1 XP_016857092.1 2, 3 238
XM_017001604.1 XP_016857093.1 2, 3 240
XM_017001608.1 XP_016857097.1 1, 2 154
XM_017001609.1 XP_016857098.1 2, 3 250
XM_017001610.1 XP_016857099.1 2, 3 240
XM_017001612.1 XP_016857101.1 1, 2 143
XM_017001613.1 XP_016857102.1 1, 2 143
XM_017001614.1 XP_016857103.1 2, 3 250
XM_017001615.1 XP_016857104.1 2, 3 250
GenBank mRNA AF272833.1 - 2, 3 184
AK001366.1 - 2, 3 235
AK056128.1 - 1, 2 327
AK092950.1 - 2, 3 235
AK222863.1 - 2, 3 232
AK290610.1 - 2, 3 281
AK297140.1 - 2, 3 235
AK309573.1 - 2, 3 227
BC002535.2 - 2, 3 251 3140498
BC070067.1 - 2, 3 258
BI911005.1 - 2, 3 268
BX537684.1 - 2, 3 258
CR749791.1 - 2, 3 232

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 2, 3 288
GenBank mRNA AK293928.1 - 2, 3 288
AK295146.1 - 2, 3 288
BC099909.1 - 2, 3 238
DC387604.1 - 2, 3 288

View Full Product Details