Gene Transcripts

Gene Symbol
GOLGA8B
Entrez Gene ID
440270
Gene Name
golgin A8 family member B
Gene Aliases
GOLGA5
Chromosome Location
Chr. 15: 34525276 - 34583651 on Build GRCh38
UniGene ID
Hs.720151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001023567.4 NP_001018861.3 16 3686
NR_027410.1 24 5934
XM_011521596.1 XP_011519898.1 23 4531
XM_011521597.1 XP_011519899.1 22 4482
XM_017022210.1 XP_016877699.1 24 4835
XM_017022211.1 XP_016877700.1 24 4604
XM_017022212.1 XP_016877701.1 23 5158
XM_017022213.1 XP_016877702.1 23 4560
XM_017022214.1 XP_016877703.1 24 4832
XM_017022215.1 XP_016877704.1 24 5357
XM_017022216.1 XP_016877705.1 23 5955
XM_017022217.1 XP_016877706.1 22 5877
XM_017022218.1 XP_016877707.1 24 5984
GenBank mRNA AL133593.1 - 1 657
BX647673.1 - 23 5902

Gene Symbol
GOLGA8A
Entrez Gene ID
23015
Gene Name
golgin A8 family member A
Gene Aliases
GM88
Chromosome Location
Chr. 15: 34379068 - 34437466 on Build GRCh38
UniGene ID
Hs.182982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181077.3 NP_851422.1 16 3684
NR_027409.1 24 5946
XM_006720440.2 XP_006720503.1 16 3649
XM_017022018.1 XP_016877507.1 20 6080
XM_017022019.1 XP_016877508.1 19 5776
XM_017022020.1 XP_016877509.1 14 3771
GenBank mRNA AB020662.1 - 15 3664
AF204231.1 - 15 4001
AL117569.1 - 2 1991
AL137686.1 - 15 3404
BC064341.1 - 1 1116
BC150329.1 - 15 3675
BC152410.1 - 15 3664
BX648160.1 - 23 5900
BX648575.1 - 23 5653

View Full Product Details