Gene Transcripts

Gene Symbol
USP22
Entrez Gene ID
23326
Gene Name
ubiquitin specific peptidase 22
Gene Aliases
USP3L
Chromosome Location
Chr. 17: 20999593 - 21043409 on Build GRCh38
UniGene ID
Hs.462492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015276.1 NP_056091.1 4 689
XM_005256575.2 XP_005256632.1 4 403
GenBank mRNA AB028986.1 - 4 689
BC110499.1 - 4 448
BC126898.1 - 4 449 40027759
BX640815.1 - 3 271

View Full Product Details