Gene Transcripts

Gene Symbol
USP24
Entrez Gene ID
23358
Gene Name
ubiquitin specific peptidase 24
Gene Aliases
Chromosome Location
Chr. 1: 55066359 - 55215374 on Build GRCh38
UniGene ID
Hs.477009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015306.2 NP_056121.2 30 3570
XM_005270690.3 XP_005270747.1 17 1611
XM_017000831.1 XP_016856320.1 31 3635
XM_017000832.1 XP_016856321.1 31 3632
XM_017000833.1 XP_016856322.1 31 3127
XM_017000834.1 XP_016856323.1 30 3067
XM_017000835.1 XP_016856324.1 31 3638
XM_017000836.1 XP_016856325.1 31 3638
XM_017000837.1 XP_016856326.1 31 3638
GenBank mRNA AK127075.1 - 13 1315

View Full Product Details