Gene Transcripts

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
Chromosome Location
Chr. 12: 105173297 - 105236230 on Build GRCh38
UniGene ID
Hs.506603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251904.1 NP_001238833.1 6 616
NM_001251905.1 NP_001238834.1 6 659
NM_018171.3 NP_060641.2 6 616
XM_006719472.1 XP_006719535.1 6 535
XM_011538530.2 XP_011536832.1 6 2068
XM_011538531.2 XP_011536833.1 6 1731
XM_011538532.2 XP_011536834.1 6 3057
XM_017019551.1 XP_016875040.1 6 1021
XM_017019552.1 XP_016875041.1 6 1731
XM_017019553.1 XP_016875042.1 7 3329
XM_017019554.1 XP_016875043.1 6 535
GenBank mRNA AK001521.2 - 6 436
AK290562.1 - 6 469
AK293783.1 - 6 408
AK296610.1 - 6 415
AK297100.1 - 6 469
AK310377.1 - 6 404
AY113704.1 - 6 616
BC033731.1 - 6 520 5168590

View Full Product Details