Gene Transcripts

Gene Symbol
TTC24
Entrez Gene ID
164118
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
Chromosome Location
Chr. 1: 156579727 - 156587717 on Build GRCh38
UniGene ID
Hs.447851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105669.2 NP_001099139.2 5, 6 1199
XM_011509253.2 XP_011507555.1 4, 5 581
XM_011509255.2 XP_011507557.1 4, 5 581
XM_017000497.1 XP_016855986.1 2, 3 203
GenBank mRNA BQ216668.1 - 3, 4 463

View Full Product Details