Gene Transcripts

Gene Symbol
ATXN1L
Entrez Gene ID
342371
Gene Name
ataxin 1 like
Gene Aliases
BOAT, BOAT1
Chromosome Location
Chr. 16: 71845991 - 71857333 on Build GRCh38
UniGene ID
Hs.743239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137675.3 NP_001131147.1 3 573
GenBank mRNA DB097328.1 - 3 575

View Full Product Details