Gene Transcripts
- Gene Symbol
- LOC105371899
- Entrez Gene ID
- 105371899
- Gene Name
- uncharacterized LOC105371899
- Gene Aliases
- Chromosome Location
- Chr. 17: 76950474 - 76969199 on Build GRCh38
- UniGene ID
- Hs.634683
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136416.1 |
|
3 |
609 |
|
GenBank mRNA |
BM700476.1 |
- |
3 |
507 |
|
CD173460.1 |
- |
3 |
383 |
|
View Full Product Details