Gene Transcripts

Gene Symbol
PRR11
Entrez Gene ID
55771
Gene Name
proline rich 11
Gene Aliases
Chromosome Location
Chr. 17: 59155499 - 59206709 on Build GRCh38
UniGene ID
Hs.631750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018304.3 NP_060774.2 5, 6 965
GenBank mRNA AK000296.1 - 2, 3 281
AK001891.1 - 5, 6 718
AK225377.1 - 5, 6 718
AK225380.1 - 5, 6 723
BC008669.1 - 5, 6 707 3867293
FJ161961.1 - 5, 6 718

View Full Product Details