Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_024430.2 | 4 | 1772 | ||
NR_137175.1 | 2 | 2043 | |||
NR_137176.1 | 2 | 1754 | |||
NR_137177.1 | 2 | 1733 | |||
NR_137179.1 | 5 | 1726 | |||
NR_137180.1 | 5 | 1784 | |||
NR_137181.1 | 3 | 1669 | |||
NR_137182.1 | 4 | 1751 | |||
NR_137183.1 | 3 | 1660 | |||
NR_137184.1 | 2 | 1578 | |||
NR_137185.1 | 3 | 1639 | |||
NR_137186.1 | 3 | 1548 | |||
NR_137187.1 | 3 | 1527 | |||
NR_137188.1 | 3 | 1617 | |||
NR_137189.1 | 2 | 2064 | |||
NR_137190.1 | 3 | 2097 | |||
NR_137191.1 | 3 | 2076 | |||
NR_137192.1 | 4 | 1936 | |||
NR_137193.1 | 5 | 1763 | |||
NR_137194.1 | 3 | 1638 | |||
NR_137195.1 | 2 | 1806 | |||
NR_137196.1 | 2 | 1827 | |||
GenBank mRNA | AB075498.1 | - | 2 | 2296 | |
AK021552.1 | - | 1 | 938 | ||
AK123947.1 | - | 2 | 1980 | ||
AL832469.1 | - | 3 | 1332 | ||
BC040599.1 | - | 2 | 1356 | ||
BX538058.1 | - | 2 | 1334 | ||
BX647608.1 | - | 2 | 1334 |