Gene Transcripts
- Gene Symbol
- C5orf22
- Entrez Gene ID
- 55322
- Gene Name
- chromosome 5 open reading frame 22
- Gene Aliases
- Chromosome Location
- Chr. 5: 31532086 - 31555058 on Build GRCh38
- UniGene ID
- Hs.519246
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018356.2 |
NP_060826.2
|
1, 2 |
204 |
|
NR_134298.1 |
|
1, 2 |
204 |
|
XM_006714479.2 |
XP_006714542.1
|
1, 2 |
178 |
|
XM_006714480.3 |
XP_006714543.1
|
1, 2 |
186 |
|
XM_011514062.2 |
XP_011512364.1
|
1, 2 |
189 |
|
XM_017009607.1 |
XP_016865096.1
|
1, 2 |
193 |
|
XM_017009608.1 |
XP_016865097.1
|
1, 2 |
204 |
|
XM_017009609.1 |
XP_016865098.1
|
1, 2 |
178 |
|
XM_017009610.1 |
XP_016865099.1
|
1, 2 |
193 |
|
XM_017009611.1 |
XP_016865100.1
|
1, 2 |
193 |
|
XM_017009612.1 |
XP_016865101.1
|
1, 2 |
194 |
|
XM_017009613.1 |
XP_016865102.1
|
1, 2 |
384 |
|
XM_017009614.1 |
XP_016865103.1
|
1, 2 |
186 |
|
XM_017009615.1 |
XP_016865104.1
|
1, 2 |
186 |
|
XM_017009616.1 |
XP_016865105.1
|
1, 2 |
193 |
|
GenBank mRNA |
AK002055.1 |
- |
1, 2 |
191 |
|
AK299156.1 |
- |
1, 2 |
181 |
|
BC021215.2 |
- |
1, 2 |
141 |
4302543 |
BC032845.1 |
- |
1, 2 |
187 |
|
DA783320.1 |
- |
1, 2 |
178 |
|
DC404356.1 |
- |
1, 2 |
204 |
|
View Full Product Details