Gene Transcripts

Gene Symbol
C22orf34
Entrez Gene ID
348645
Gene Name
chromosome 22 open reading frame 34
Gene Aliases
Chromosome Location
Chr. 22: 49414524 - 49657542 on Build GRCh38
UniGene ID
Hs.133159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110522.1 3 996
NR_110523.1 5 1467
GenBank mRNA BC033837.1 - 5 1440

View Full Product Details