Gene Transcripts

Gene Symbol
TMEM242
Entrez Gene ID
729515
Gene Name
transmembrane protein 242
Gene Aliases
BM033, C6orf35
Chromosome Location
Chr. 6: 157289022 - 157324259 on Build GRCh38
UniGene ID
Hs.157212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018452.5 NP_060922.2 1, 2 856
GenBank mRNA AF161405.1 - 1, 2 193
AF217510.1 - 1, 2 198
AK000891.1 - 1, 2 104
BC029130.1 - 1, 2 124 5182086
BC146893.1 - 1, 2 166 9056821
BC146895.1 - 1, 2 166 9056823
DA684766.1 - 1, 2 359

View Full Product Details