Gene Transcripts

Gene Symbol
ITFG2
Entrez Gene ID
55846
Gene Name
integrin alpha FG-GAP repeat containing 2
Gene Aliases
MDS028
Chromosome Location
Chr. 12: 2812621 - 2832217 on Build GRCh38
UniGene ID
Hs.446098
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018463.3 NP_060933.3 3, 4 377
NR_130744.1 3, 4 376
XM_005253707.4 XP_005253764.1 3, 4 346
GenBank mRNA AF220048.1 - 631
AK027483.1 - 3, 4 301
AK130319.1 - 2, 3 907
AK291110.1 - 3, 4 305
AK300905.1 - 3, 4 305
AY599883.1 - 3, 4 237
BC006552.2 - 3, 4 262 3029909
BC013399.2 - 3, 4 262 3030623
BI261823.1 - 3, 4 377
BQ058412.1 - 376
KU178719.1 - 3, 4 237
KU178720.1 - 3, 4 237
KU178721.1 - 3, 4 237

View Full Product Details