Gene Transcripts

Gene Symbol
LRRC27
Entrez Gene ID
80313
Gene Name
leucine rich repeat containing 27
Gene Aliases
Chromosome Location
Chr. 10: 132330063 - 132381506 on Build GRCh38
UniGene ID
Hs.119897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143757.1 NP_001137229.1 11 7595
NM_030626.2 NP_085129.1 11 7442
GenBank mRNA AL512722.1 - 1 1876

View Full Product Details