Gene Transcripts

Gene Symbol
LOC440934
Entrez Gene ID
440934
Gene Name
uncharacterized LOC440934
Gene Aliases
Chromosome Location
Chr. 2: 222318471 - 222357910 on Build GRCh38
UniGene ID
Hs.732732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136638.1 2 1126
NR_136639.1 2 830
NR_136640.1 2 395
NR_136641.1 2 574
GenBank mRNA BC008048.2 - 2 1125

View Full Product Details