Gene Transcripts

Gene Symbol
ING5
Entrez Gene ID
84289
Gene Name
inhibitor of growth family member 5
Gene Aliases
p28ING5
Chromosome Location
Chr. 2: 241687017 - 241729481 on Build GRCh38
UniGene ID
Hs.529172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017005100.1 XP_016860589.1 1 423
XM_017005101.1 XP_016860590.1 1 423

View Full Product Details