Gene Transcripts

Gene Symbol
KIAA1257
Entrez Gene ID
57501
Gene Name
KIAA1257
Gene Aliases
Chromosome Location
Chr. 3: 128910186 - 129032431 on Build GRCh38
UniGene ID
Hs.518247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020741.2 NP_065792.1 8 1578
GenBank mRNA AB033083.1 - 8 1550
BC031632.1 - 7 1539 5170293
BC038736.1 - 8 1511

View Full Product Details