Gene Transcripts

Gene Symbol
RBM44
Entrez Gene ID
375316
Gene Name
RNA binding motif protein 44
Gene Aliases
Chromosome Location
Chr. 2: 237798048 - 237842808 on Build GRCh38
UniGene ID
Hs.607265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080504.2 NP_001073973.2 9 2394
XM_005246075.1 XP_005246132.1 9 2735
XM_011511152.2 XP_011509454.1 9 2735
XM_011511153.2 XP_011509455.1 9 2735
XM_017004054.1 XP_016859543.1 9 2735
XM_017004055.1 XP_016859544.1 9 3105
XM_017004056.1 XP_016859545.1 9 2675
XM_017004057.1 XP_016859546.1 9 2735
XM_017004058.1 XP_016859547.1 4 1073
GenBank mRNA BC126111.1 - 6 477
BX648107.1 - 8 3047

View Full Product Details