Gene Transcripts

Gene Symbol
TTC24
Entrez Gene ID
164118
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
Chromosome Location
Chr. 1: 156579727 - 156587717 on Build GRCh38
UniGene ID
Hs.447851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105669.2 NP_001099139.2 9 1515
XM_011509253.2 XP_011507555.1 8 897
XM_011509254.2 XP_011507556.1 8 894
XM_017000497.1 XP_016855986.1 6 519

View Full Product Details