Gene Transcripts

Gene Symbol
TBC1D12
Entrez Gene ID
23232
Gene Name
TBC1 domain family member 12
Gene Aliases
Chromosome Location
Chr. 10: 94402429 - 94536332 on Build GRCh38
UniGene ID
Hs.500598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015188.1 NP_056003.1 3 1343
XM_006717732.3 XP_006717795.2 3 1343
XM_006717733.2 XP_006717796.1 4 303
XM_011539558.2 XP_011537860.1 3 354
XM_017015996.1 XP_016871485.1 3 1343
XM_017015997.1 XP_016871486.1 3 1343
XM_017015998.1 XP_016871487.1 2 130
XM_017015999.1 XP_016871488.1 2 346
GenBank mRNA AB011180.1 - 3 1159
AB449898.1 - 3 1140
AL049956.1 - 2 91

View Full Product Details