Gene Transcripts

Gene Symbol
SLC4A8
Entrez Gene ID
9498
Gene Name
solute carrier family 4 member 8
Gene Aliases
NBC3, NDCBE
Chromosome Location
Chr. 12: 51391317 - 51515763 on Build GRCh38
UniGene ID
Hs.4749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039960.2 NP_001035049.1 25 4161
NM_001258401.2 NP_001245330.1 25 4107
XM_011539010.1 XP_011537312.1 26 4216
XM_011539011.1 XP_011537313.1 26 4161
XM_011539014.2 XP_011537316.1 16 2814
XM_017020240.1 XP_016875729.1 26 4243
XM_017020242.1 XP_016875731.1 26 4230
GenBank mRNA AK093005.1 - 1 823
AK093357.1 - 4 1453
AK310017.1 - 6 1284
DQ063579.2 - 25 4150
DQ975204.1 - 25 3977

View Full Product Details