Gene Transcripts
- Gene Symbol
- SLC4A8
- Entrez Gene ID
- 9498
- Gene Name
- solute carrier family 4 member 8
- Gene Aliases
- NBC3, NDCBE
- Chromosome Location
- Chr. 12: 51391317 - 51515763 on Build GRCh38
- UniGene ID
- Hs.4749
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039960.2 |
NP_001035049.1
|
24, 25 |
3447 |
|
NM_001258401.2 |
NP_001245330.1
|
24, 25 |
3393 |
|
XM_011539010.1 |
XP_011537312.1
|
25, 26 |
3502 |
|
XM_011539011.1 |
XP_011537313.1
|
25, 26 |
3447 |
|
XM_011539014.2 |
XP_011537316.1
|
15, 16 |
2100 |
|
XM_017020240.1 |
XP_016875729.1
|
25, 26 |
3529 |
|
XM_017020242.1 |
XP_016875731.1
|
25, 26 |
3516 |
|
GenBank mRNA |
AK093357.1 |
- |
3, 4 |
739 |
|
AK310017.1 |
- |
5, 6 |
570 |
|
DQ063579.2 |
- |
24, 25 |
3436 |
|
DQ975204.1 |
- |
24, 25 |
3263 |
|
View Full Product Details