Gene Transcripts

Gene Symbol
PCDHB2
Entrez Gene ID
56133
Gene Name
protocadherin beta 2
Gene Aliases
PCDH-BETA2
Chromosome Location
Chr. 5: 141094607 - 141097380 on Build GRCh38
UniGene ID
Hs.533023
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018936.3 NP_061759.1 1 519
GenBank mRNA AB209745.1 - 1 295
AF152495.1 - 1 335
AF217756.1 - 1 335
AK027526.1 - 1 491
AK314642.1 - 1 473
BC098347.1 - 1 382 40007951
BC098575.1 - 1 483 30921306
BF206341.1 - 1 472
DA703023.1 - 1 519

View Full Product Details