Gene Transcripts

Gene Symbol
PCDHB6
Entrez Gene ID
56130
Gene Name
protocadherin beta 6
Gene Aliases
PCDH-BETA6
Chromosome Location
Chr. 5: 141150022 - 141153287 on Build GRCh38
UniGene ID
Hs.283085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018939.3 NP_061762.2 1 449
GenBank mRNA AF152499.1 - 1 213
AF217752.1 - 1 213
AK313482.1 - 1 370
DA789782.1 - 1 449

View Full Product Details