Gene Transcripts

Gene Symbol
SH3BP1
Entrez Gene ID
23616
Gene Name
SH3 domain binding protein 1
Gene Aliases
ARHGAP43
Chromosome Location
Chr. 22: 37639677 - 37656043 on Build GRCh38
UniGene ID
Hs.601143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018957.3 NP_061830.3 18 2385
GenBank mRNA AL157480.1 - 7 1209
BC008282.1 - 16 2178 3639782

View Full Product Details