Gene Transcripts

Gene Symbol
ZFAS1
Entrez Gene ID
441951
Gene Name
ZNFX1 antisense RNA 1
Gene Aliases
C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1
Chromosome Location
Chr. 20: 49278178 - 49289260 on Build GRCh38
UniGene ID
Hs.356766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003604.2 1 453
NR_003606.2 1 453
NR_036658.1 1 453
GenBank mRNA AY513722.1 - 1 453
BM706184.1 - 1 453

View Full Product Details