Gene Transcripts

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
Chromosome Location
Chr. 10: 87094478 - 87191468 on Build GRCh38
UniGene ID
Hs.500419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019054.2 NP_061927.2 3 812
XM_006717903.3 XP_006717966.1 3 802
XM_006717905.3 XP_006717968.1 2 725
XM_011539876.2 XP_011538178.1 1 703
XM_011539877.2 XP_011538179.1 4 867
XM_011539878.2 XP_011538180.1 2 725
XM_011539879.2 XP_011538181.1 1 711
XM_011539881.2 XP_011538183.1 1 703
XM_011539882.2 XP_011538184.1 1 703
XM_017016349.1 XP_016871838.1 6 1111
XM_017016350.1 XP_016871839.1 3 1070
XM_017016351.1 XP_016871840.1 3 802
XM_017016352.1 XP_016871841.1 3 802
GenBank mRNA AK022978.1 - 3 812
BC051863.2 - 2 758 6146593

Gene Symbol
FAM35BP
Entrez Gene ID
414241
Gene Name
family with sequence similarity 35, member A pseudogene
Gene Aliases
FAM35B, FAM35EP, bA38L15.1
Chromosome Location
Chr. 10: 46610472 - 46651976 on Build GRCh38
UniGene ID
Hs.594331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027632.1 1 704

Gene Symbol
FAM35DP
Entrez Gene ID
439965
Gene Name
family with sequence similarity 35, member A pseudogene
Gene Aliases
FAM35B2
Chromosome Location
Chr. 10: 47689700 - 47731218 on Build GRCh38
UniGene ID
Hs.283521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027634.1 1 704

View Full Product Details