Gene Transcripts

Gene Symbol
CT45A10
Entrez Gene ID
102723631
Gene Name
cancer/testis antigen family 45, member A10
Gene Aliases
Chromosome Location
Chr. X: 135880752 - 135893577 on Build GRCh38
UniGene ID
Hs.535081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291527.1 NP_001278456.1 3 495
NM_001291528.1 NP_001278457.1 3 478
NM_001291529.1 NP_001278458.1 3 416
NM_001291530.1 NP_001278459.1 3 433
XM_011531238.2 XP_011529540.1 3 429
XM_011531239.1 XP_011529541.1 3 410
GenBank mRNA AK292207.1 - 3 412
AY743713.1 - 3 571
BC070138.1 - 3 416 4685945
BC144429.1 - 3 470 9052950
BC144430.1 - 3 487 9052951
BC144527.1 - 3 493 9053054
BC144528.1 - 3 478 9053055

View Full Product Details