Gene Transcripts
- Gene Symbol
- CT45A10
- Entrez Gene ID
- 102723631
- Gene Name
- cancer/testis antigen family 45, member A10
- Gene Aliases
- Chromosome Location
- Chr. X: 135880752 - 135893577 on Build GRCh38
- UniGene ID
- Hs.535081
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY743713.1 |
- |
3 |
539 |
|
BC070138.1 |
- |
3 |
384 |
4685945 |
View Full Product Details