Gene Transcripts

Gene Symbol
SCGB2B2
Entrez Gene ID
284402
Gene Name
secretoglobin family 2B member 2
Gene Aliases
SCGB4A2, SCGBL
Chromosome Location
Chr. 19: 34590625 - 34594585 on Build GRCh38
UniGene ID
Hs.656990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025591.3 NP_001020762.1 2 165
GenBank mRNA AK093495.1 - 3 2230
BC093909.1 - 2 165 7939754
BC093935.1 - 2 165 7939780

View Full Product Details