Gene Transcripts

Gene Symbol
MME
Entrez Gene ID
4311
Gene Name
membrane metallo-endopeptidase
Gene Aliases
CALLA, CD10, CMT2T, NEP, SFE
Chromosome Location
Chr. 3: 155024124 - 155183729 on Build GRCh38
UniGene ID
Hs.307734
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000902.3 NP_000893.2 4 396
NM_007287.2 NP_009218.2 4 372
NM_007288.2 NP_009219.2 4 418
NM_007289.2 NP_009220.2 4 463
XM_006713646.3 XP_006713709.1 4 410
XM_006713647.3 XP_006713710.1 4 3556
XM_011512855.2 XP_011511157.1 4 487
XM_011512856.2 XP_011511158.1 4 414
XM_011512857.2 XP_011511159.1 4 385
GenBank mRNA AK291761.1 - 4 487
BC101632.1 - 4 354 8069138
BC101658.1 - 4 352 8069164
BC143465.1 - 4 354 9051973
BM151602.1 - 4 396
BX647134.1 - 4 368
DB142029.1 - 4 414
J03779.1 - 3 287
X07166.1 - 3 272
Y00811.1 - 4 411

View Full Product Details