Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020165.3 | NP_064550.3 | 6, 7 | 788 | |
XM_017006873.1 | XP_016862362.1 | 7, 8 | 946 | ||
GenBank mRNA | AB035274.2 | - | 6, 7 | 823 | |
AF169796.1 | - | 6, 7 | 768 | ||
AK023075.1 | - | 6, 7 | 775 | ||
AK222771.1 | - | 6, 7 | 766 | ||
AY004333.1 | - | 6, 7 | 734 | ||
BC001302.1 | - | 6, 7 | 755 | 3451960 |