Gene Transcripts

Gene Symbol
SLC10A3
Entrez Gene ID
8273
Gene Name
solute carrier family 10 member 3
Gene Aliases
DXS253E, P3
Chromosome Location
Chr. X: 154487306 - 154490690 on Build GRCh38
UniGene ID
Hs.522826
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142391.2 NP_001135863.1 3 638
NM_001142392.2 NP_001135864.1 3 638
NM_019848.4 NP_062822.1 2 873
XM_005277913.4 XP_005277970.1 2 1295
XM_006724847.3 XP_006724910.1 2 816
XM_006724848.3 XP_006724911.1 2 816
XM_011531200.2 XP_011529502.2 2 848
XM_011531201.2 XP_011529503.1 2 552
GenBank mRNA BC004966.1 - 3 562
BX401493.2 - 3 610

View Full Product Details