Gene Transcripts

Gene Symbol
SAR1A
Entrez Gene ID
56681
Gene Name
secretion associated Ras related GTPase 1A
Gene Aliases
SAR1, SARA1, Sara, masra2
Chromosome Location
Chr. 10: 70150204 - 70170529 on Build GRCh38
UniGene ID
Hs.499960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142648.1 NP_001136120.1 8 894
NM_020150.4 NP_064535.1 7 824
GenBank mRNA AF087897.1 - 7 791
AF217959.1 - 7 791
AF274026.1 - 7 790
AF318066.1 - 6 690
AK298591.1 - 6 657
AY008268.1 - 7 815
BC003658.2 - 7 789 3546972

View Full Product Details