Gene Transcripts

Gene Symbol
FEM1C
Entrez Gene ID
56929
Gene Name
fem-1 homolog C
Gene Aliases
EUROIMAGE686608, EUROIMAGE783647, FEM1A
Chromosome Location
Chr. 5: 115520908 - 115544919 on Build GRCh38
UniGene ID
Hs.47367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020177.2 NP_064562.1 2 719
XM_005272035.4 XP_005272092.1 2 721
XM_017009647.1 XP_016865136.1 2 744
GenBank mRNA AB058688.1 - 1 247
AF391093.1 - 1 157
AK315803.1 - 2 497
AL365415.1 - 2 457
AY249188.1 - 1 157
BC028369.2 - 2 721 4826273

View Full Product Details