Gene Transcripts

Gene Symbol
Map2k4
Entrez Gene ID
287398
Gene Name
mitogen activated protein kinase kinase 4
Gene Aliases
MKK4, Sek1
Chromosome Location
Chr. 10: 52196121 - 52301887 on Build Rnor_6.0
UniGene ID
Rn.198875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030023.1 NP_001025194.1 5 645
GenBank mRNA AY880882.1 - 5 645
KC795688.1 - 6 678

View Full Product Details