Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033888.1 | NP_001029060.1 | 8 | 943 | |
XM_006228565.2 | XP_006228627.1 | 7 | 4110 | ||
XM_006228566.2 | XP_006228628.1 | 7 | 4107 | ||
XM_006228567.2 | XP_006228629.1 | 8 | 1169 | ||
XM_006228568.2 | XP_006228630.1 | 8 | 1375 | ||
GenBank mRNA | AB210107.1 | - | 8 | 943 | |
BC129073.1 | - | 8 | 1056 | 7189989 |