Gene Transcripts

Gene Symbol
Apopt1
Entrez Gene ID
299341
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
APOP-1, Apop1, RGD1304719
Chromosome Location
Chr. 6: 136185772 - 136210936 on Build Rnor_6.0
UniGene ID
Rn.3813
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037769.2 NP_001032858.1 261
GenBank mRNA BC107661.1 - 256

View Full Product Details