Gene Transcripts

Gene Symbol
Adipor2
Entrez Gene ID
312670
Gene Name
adiponectin receptor 2
Gene Aliases
Chromosome Location
Chr. 4: 151412135 - 151480108 on Build Rnor_6.0
UniGene ID
Rn.101984
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037979.1 NP_001033068.1 7 1066
XM_006237183.1 XP_006237245.1 8 1372
XM_006237184.2 XP_006237246.1 8 1243
XM_006237185.2 XP_006237247.1 8 1258
XM_006237186.2 XP_006237248.1 8 1387
GenBank mRNA AY724514.1 - 8 1326
DQ148392.1 - 7 1066
FQ232508.1 - 8 1345

View Full Product Details