Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020165.3 | NP_064550.3 | 9 | 1097 | |
XM_017006873.1 | XP_016862362.1 | 10 | 1255 | ||
GenBank mRNA | AB035274.2 | - | 9 | 1132 | |
AF169796.1 | - | 9 | 1077 | ||
AK023075.1 | - | 9 | 1084 | ||
AK222771.1 | - | 9 | 1075 | ||
AY004333.1 | - | 9 | 1043 | ||
BC001302.1 | - | 9 | 1064 | 3451960 |