Gene Transcripts

Gene Symbol
Olfr1463
Entrez Gene ID
258120
Gene Name
olfactory receptor 1463
Gene Aliases
MOR202-29P
Chromosome Location
Chr. 19: 13234252 - 13235184 on Build GRCm38
UniGene ID
Mm.329753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011840.1 NP_001011840.1 1 15

View Full Product Details