Gene Transcripts

Gene Symbol
Rcbtb1
Entrez Gene ID
71330
Gene Name
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1
Gene Aliases
5430409I18Rik, AW111883, CLLD7, CLLL7
Chromosome Location
Chr. 14: 59201027 - 59237267 on Build GRCm38
UniGene ID
Mm.234195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027764.2 NP_082040.2 12 2762
GenBank mRNA AK088299.1 - 12 2751
BC040764.1 - 5 1457
BC067005.1 - 13 2816 5721370

View Full Product Details