Gene Transcripts

Gene Symbol
4921506M07Rik
Entrez Gene ID
70846
Gene Name
RIKEN cDNA 4921506M07 gene
Gene Aliases
Chromosome Location
Chr. 12: 57564115 - 57622105 on Build GRCm38
UniGene ID
Mm.449996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037743.1 NP_001032832.1 3 1055
GenBank mRNA AK014828.1 - 3 1046
BC100489.1 - 3 1055 30915676

View Full Product Details