Gene Transcripts

Gene Symbol
Pih1d3
Entrez Gene ID
74708
Gene Name
PIH1 domain containing 3
Gene Aliases
4930521A18Rik
Chromosome Location
Chr. 1: 31222839 - 31224287 on Build GRCm38
UniGene ID
Mm.246796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029062.2 NP_083338.1 1 130
GenBank mRNA AK015858.1 - 1 131
BC107262.2 - 1 122 40052516

View Full Product Details