Gene Transcripts

Gene Symbol
Cln6
Entrez Gene ID
76524
Gene Name
ceroid-lipofuscinosis, neuronal 6
Gene Aliases
1810065L06Rik, AW743417, D9Bwg1455e, nclf
Chromosome Location
Chr. 9: 62838787 - 62852002 on Build GRCm38
UniGene ID
Mm.283636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033175.2 NP_001028347.1 7 1911
GenBank mRNA AK154414.1 - 7 1912
AK154551.1 - 7 1907
AK207918.1 - 1 345
AK215418.1 - 1 346

View Full Product Details