Gene Transcripts

Gene Symbol
HYKK
Entrez Gene ID
123688
Gene Name
hydroxylysine kinase
Gene Aliases
AGPHD1
Chromosome Location
Chr. 15: 78507564 - 78537373 on Build GRCh38
UniGene ID
Hs.307962
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK126453.1 - 5 1127
BC132753.1 - 4 906 40146775
BC144383.1 - 4 906 9052904

View Full Product Details